missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ TRACKMAN Connected Starter Kit, Multi Channel

The TRACKMAN Connected Starter Kit includes the pipette models that match with your QIAGEN protocols. When pairing TRACKMAN Connected with PIPETMAN™ M Connected, the installed app guides you through your QIAGEN-programmed protocol while automatically adjusting the pipette volume settings.

Brand:  Gilson™ FB10211


4483.14 GBP valid until 2024-12-31
Use promo code "23279" to get your promotional price.



Additional Details : Weight : 6.63300kg

 View more versions of this product

Product Code. 17379243

  • £4810.00 / 1 set
Estimated Shipment: 10-09-2024
to see stock.

Description

Description

We've partnered with QIAGEN, whose well-known kits help your molecular biology applications. You can now execute your QIAGEN protocols with guidance from the TRACKMAN™ Connected pipetting system. Our pipettes and tips match perfectly with the volumes needed to start your protocol, all available in one unique kit.

The TRACKMAN Connected Starter Kit Multichannel includes the following:

  • TRACKMAN Connected tablet and accessories
  • PIPETMAN™ M Connected P1200M
  • PIPETMAN™ M Connected P8x200M
  • PIPETMAN™ M Connected P8x1200M
  • 10 racks of 1200 μL filter tips
  • 10 racks of 200 μL filter tips

Optimize your protocol execution with TRACKMAN Connected Starter Kit Multichannel and QIAGEN kits for the purification or extraction of RNA virus.

  • NOTE: All QIAGEN kits are sold separately and must be purchased through QIAGEN directly

Our TRACKMAN Connected Starter Kit Multichannel is compatible with:

  • QIAamp 96 Viral RNA Kit: Viral RNA purification from cell-free body fluids in 96-well format. Suitable for SARS-CoV-2 detection.
Specifications

Specifications

1200 μL, 200 μL tips
Not Autoclavable
Variable
All Pipettes with Tips Mounted
Polypropylene
8
Yes
Pipette Controller
Pipette tips
Certified Quality
No
Assorted
Reusable
Ergonomic
TRACKMAN Connected
0.45 μm
Non-sterile
1200 μL
UK-Plug
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers