missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Series CB - CO2 Incubator, with Hot Air Sterilization and Heat Sterilizable CO2 Sensor

The powerful and versatile CB CO2 incubator is suitable for cell-based assays in drug and medical research. It is even suitable for complex cultivation experiments or individual environments with hypoxic conditions thanks to its comprehensive assortment of options and accessories.

Brand:  BINDER™ 9640-0005


6409.53 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.



Additional Details : Weight : 54.00000kg

 View more versions of this product

Product Code. 17383693

  • £7915.00 / Each
Estimated Shipment: 29-07-2024
to see stock.

Description

Description

  • Optimized double-pan humidification system with condensation protection
  • Tried-and-tested anti-contamination concept with 180°C hot air sterilization
  • Hot-air sterilizable CO2 sensor with single-beam infrared technology
  • Seamless inner chamber made of stainless steel with flanges as shelf support system
  • Fanless interior with Venturi CO2 gas mixing nozzle
  • Intuitive touchscreen controller
  • Internal data recording, can be read out in open format via USB interface
  • Fault diagnosis system with visual and acoustic alarm
  • Interfaces: Ethernet, USB, zero-voltage alarm contact
  • Fail-safe CO2 system for protecting the pH of cell cultures
Specifications

Specifications

1.9 cu. ft.
10 kg
53 L
0 to 20%
Fanless interior
None
Stainless Steel
2
None
Ambient +4° to 60°C
TFT touch
IR sensor
400 x 330 x 400 mm
53 kg
0.8 kW
50/60 Hz
22 lb.
CB056-230 V, Standard, Series CB - CO2 incubators with hot air sterilization and heat sterilizable CO2 sensor, 53 L, 230 V 1 ∼ 50/60 Hz
Stainless Steel
IR Sensor
Natural
No Lighting
1
1 to 21%
Nema 5-20P
CO2 Incubator
CB056-230 V
IR sensor
528 x 645 x 674 mm
230 V
3.5 A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Services