Learn More
Thermo Scientific™ Productainer™ 3D BioProcess Container (BPC), Aegis™ 5-14 film, four ports, Luer locks + MPX
Thermo Scientific Productainer 3D BioProcess Containers (BPCs) are used to package, store, and transport media, buffers, water for injection (WFI), process liquids, and biological products.
£1545.00
Specifications
Description | Top Dispense |
---|---|
Film Description | Aegis 5-14 |
Port Detail | 4 ports |
Tubing Style | C-Flex |
Description
Productainer BPCs are constructed with Aegis 5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainer BPCs are suitable for a wide array of bioprocess applications. For optimal fit, form, and function, Productainer BPCs can be customized using the industry's largest component library.
Highlights:
• Film optimized for flexibility and durability—superior stress resistance, puncture resistance, and physical barrier properties to ensure stability of contents
• Gamma-irradiation sterilized
• Squared tube design with polyethylene ports located on the top of the BPC for easy access
• Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
• Customizable via the industry's largest library of qualified components
Recommended for:
• Harvest—collection and storage of bulk product from bioreactors
• Storage—sterile storage of media, buffers, and process intermediates
• Separation—feeding and receiving of sterile liquids from separation systems
• Purification—buffer feeding to purification system, fraction collection and storage
• Filling—sterile reservoir for filling systems
• Transport and shipping—bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
Top Dispense | |
Aegis 5-14 | |
4 ports | |
C-Flex |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.