Learn More
Thermo Scientific™ Productainer™ BPC Bag with 3 Top and 1 Bottom Ports
Using these BPC bags with 3 top and 1 bottom ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.
Brand: Thermo Scientific™ SH30652.01
Additional Details : Weight : 1.00000kg
Description
Using Thermo Scientific™ Productainer™ BPC bag with 3 top and 1 bottom ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.
Productainer BPCs are constructed with our Thermo Scientific CX5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Productainers can be customized using the industry's largest component library.Highlights:
- Three sizes - 50, 100 and 200L
- Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
- Gamma-irradiation sterilized
- Squared tube design with polyethylene ports located on the top of the BPC for easy access
- Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
- Customizable via the industry's largest library of qualified components
Recommended for:
Harvest - collection and storage of bulk product from bioreactors
Storage - sterile storage of media, buffers and process intermediates
Separation - Feeding and receiving of sterile liquids from separation system
Purification - Buffer feeding to purification system, fraction collection and storage
Filling - Sterile reservoir for filling systems
Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
Ambient | |
50 L | |
Packages are placed in strong cardboard cartons with product and lot identification. | |
Liners are packaged and sealed in two independent outer layers while still in the Class 10,000/ISO Class 7 area. | |
No |
Three-dimensional disposable cell culture/bioprocess bag | |
CX3-9 Film | |
3/8 in. Quick Disconnect Insert (MPC Series); 1/4 in. Male Luer Lock; 1/4 in. Female Luer Lock; 3/8 in. Quick Disconnect Body (MPC Series) | |
Gamma Irradiated |