Learn More
Kimberly-Clark Professional™ White Luxury Foam Hand Cleanser Dispenser
Ideal for the smallest of washrooms. Kimberly-Clark™ White Luxury Foam Hand Cleanser Dispenser is a compact hand hygiene bundle that provides a space saving solution.
Brand: Kimberly-Clark Professional™ 6982
28.75 GBP valid until 2024-12-31
Use promo code "22916" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Delivers the perfect amount of soap for one hand wash each time
- High capacity foam with up to 500 shots per re-fill
- Easy to keep clean with no dirt or dust traps
Specifications
Hand Cleanser Dispenser | |
19 x 8.5 x 6.5 cm (L x W x H) | |
6.5 cm | |
Ideal for use in hygiene-critical washrooms where there’s less wall space. |
White | |
8.5 cm | |
200 mL | |
1 Ea. |