Learn More
Uvex™ ProfiErgo ENB 20 Nitrile-Coated Assembly Gloves
Click to view available options
Size:
6
7
8
9
Description
- Thick, special NBR coating is resistant to oil and grease
- High-quality cotton interlock liner is comfortable, feels pleasant against the skin, and aids in regulation of the glove climate
- Enhanced grip capabilities in damp, wet, and oily areas
- Gloves are suitable for operating touch displays
- Ideal for a variety of applications including assembly, control work, and maintenance
Specifications
Specifications
Coating Material | NBR |
Color | White, Yellow |
Size | 6 |
EN388 Standard | 2121X |
Glove Cuff Style | Knit |
Liner Material | Interlock Cotton |
Series | ProfiErgo ENB 20 |