missing translation for 'onlineSavingsMsg'
Learn More

Ansell Profile 3000 Nitrile Examination Gloves

Product Code. 12950825
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 100
Pack of 1000
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 12950825

Brand: Ansell PNA3105XL

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Combine comfort and chemical resistance with Nitritex™ Profile 3000 nitrile exam gloves. Ambidextrous, latex-free glove offers chemical and puncture resistance.

  • Nitrile is resistant to a wide range of chemicals
  • Excellent puncture resistance
  • Beaded cuff provides easier donning and stability
  • Textured surface for increased grip
  • Powder-free and latex-free to eliminate Type I latex allergies
  • Wrist-length gloves measure 240mm/9.5 in.
  • Easy-access 100-glove dispenser box
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 9001:2008, 13485:2003 and ISO 2859
Classification Class I Medical Device
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Large
Sterility Non-sterile
For Use With (Application) Premium examinations
Glove Cuff Style Beaded/Rolled
Inspected for Medical Use Yes
Model Profile 3000
Packaging Type Boxed
Quantity 100/Pk.
Series Profile 3000
Length (Metric) 24 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.