missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ WTW™ ProfiLine™ pH Portable Meters
Obtain optimum results with this easy and robust portable pH/mV meter with an automatic autoread function for routine pH measurements that is safe, secure, reliable, and prevents errors.
£731.00
Specifications
Accuracy (mV) | ±0.3, ±1 |
---|---|
Accuracy (pH) | ±0.1, ±0.01, ±0.005 |
Accuracy (Temperature) | ±0.1°C |
For Use With (Application) | Ideal for all seeking an easy, robust and waterproof instrument for portable pH measuring |
Range (pH) | -2 to 19.999 |
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
10703774
|
WTW™
2AA112 |
Carry case with pH electrode SenTix™ 41, Buffer solution STP 4 and STP 7, stand, beaker, short instruction manual, CD-ROM and batteries |
£731.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||||
Description
- Clear keypad with only 6 keys
- Automatic AutoRead function for repeatable measured values make pH measurement safe and prevent errors
- Anti-skidding keypad that can be operated with gloves
- Large display that is clear and easy to read
- Easy and robust portable pH/mV meter for routine measurement
- Secure and reliable pH measurement due to repeatable results
- Waterproof according IP 67, 3-point calibration, integrated calibration timer
Specifications
±0.3, ±1 | |
±0.1°C | |
-2 to 19.999 | |
-1200 to 1200, -2000 to 2000 | |
1, 2 or 3 Calibration WTW Technical Buffer, DIN/NIST buffers | |
CE | |
80 W x 180 H mm | |
3 Years | |
All values ±1 digit | |
AA, NiMH | |
2500 hr. | |
80 mm | |
55 mm |
±0.1, ±0.01, ±0.005 | |
Ideal for all seeking an easy, robust and waterproof instrument for portable pH measuring | |
±0.005 | |
±0.3, ±1mV | |
LCD, 7 Segment | |
IP-67 | |
0.6 kg | |
4 x 1.5V AA or 4 x 1.2V NiMH rechargeable | |
-5°C to +105°C | |
Portable pH meter | |
80 W mm | |
180 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ WTW™ ProfiLine™ pH Portable Meters