missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone Receptor Antibody Cocktail, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA112626
This item is not returnable.
View return policy
Description
MA1-12626 detects Progesterone Receptor in human, equine and porcine samples. MA1-12626 has been successfully used in immunofluorescence, immunoprecipitation, western blot and immunohistochemistry (paraffin, frozen) procedures. The MA1-12626 immunogen is human endometrial carcinoma from in athymic mice.
The progesterone receptor (PR) is a member of the steroid family of nuclear receptors. The PR mediates the physiological effects of progesterone, which plays a central role in reproductive events associated with the establishment and maintenance of pregnancy. PR is found as a 94 kDa protein (Form A) or a 120 kDa protein (Form B) due to the use of alternative translation initiation sites. PR-B is the transcriptionally active form and is responsible for activating genes for the maintenance of the endometrium, maintenance of pregnancy, and inhibition of ovulation. PR-A is identical to PR-B except for a 165 amino acid deletion at the N-terminus. This deletion exposes a 140 amino acid inhibitory domain (ID) that acts as a repressor of steroid hormone transcriptional activity. In its inactive state, PgR forms a multiprotein complex which includes heat shock proteins and immunophins. Upon binding of progesterone hormone to its receptor, there is a conformational change that allows dimerization and binding of the receptor to progesterone response elements (PRE) sequences, resulting in activated transcription. A Null mutation in the PGR gene leads to pleiotrophic reproductive abnormalities.Specifications
Progesterone Receptor | |
Cocktail | |
1 mg/mL | |
PBS with no preservative | |
P06401 | |
PGR | |
Human endometrial carcinoma from in athymic mice. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ChIP Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
hPRa2, hPRa3 | |
Unconjugated | |
PGR | |
9930019P03Rik; BB114106; ENSMUSG00000074510; LOW QUALITY PROTEIN: progesterone receptor; NR3C3; Nuclear receptor subfamily 3 group C member 3; p4 receptor; PGR; pot. alt. progesterone receptor; putative; PR; PR-A; PR-B; Progesterone receptor; progesterone receptor A; progesterone-receptor; putative form B; steroid hormone receptor; testicular progesterone receptor; unnamed protein product | |
Mouse | |
Protein G | |
RUO | |
5241 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |