missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone Receptor Monoclonal Antibody (SP2), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA514505
This item is not returnable.
View return policy
Description
MA5-14505 targets Progesterone Receptor in IHC (P) and WB applications and shows reactivity with Human samples. MA5-14505 has been successfully used in western blotting, immunofluorescence, and immunohistochemistry analysis of Progesterone Receptor in human and mouse samples. The MA5-14505 immunogen is recombinant protein encoding human progesterone receptor 412-526 aa.
The progesterone receptor (PR) is a member of the steroid family of nuclear receptors. The PR mediates the physiological effects of progesterone, which plays a central role in reproductive events associated with the establishment and maintenance of pregnancy. PR is found as a 94 kDa protein (Form A) or a 120 kDa protein (Form B) due to the use of alternative translation initiation sites. PR-B is the transcriptionally active form and is responsible for activating genes for the maintenance of the endometrium, maintenance of pregnancy, and inhibition of ovulation. PR-A is identical to PR-B except for a 165 amino acid deletion at the N-terminus. This deletion exposes a 140 amino acid inhibitory domain (ID) that acts as a repressor of steroid hormone transcriptional activity. In its inactive state, PgR forms a multiprotein complex which includes heat shock proteins and immunophins. Upon binding of progesterone hormone to its receptor, there is a conformational change that allows dimerization and binding of the receptor to progesterone response elements (PRE) sequences, resulting in activated transcription. A Null mutation in the PGR gene leads to pleiotrophic reproductive abnormalities.Specifications
Progesterone Receptor | |
Monoclonal | |
0.08 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.2 | |
P06401 | |
PGR | |
Recombinant protein encoding human progesterone receptor 412-526 aa | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
SP2 | |
Unconjugated | |
PGR | |
9930019P03Rik; BB114106; ENSMUSG00000074510; LOW QUALITY PROTEIN: progesterone receptor; NR3C3; Nuclear receptor subfamily 3 group C member 3; p4 receptor; PGR; pot. alt. progesterone receptor; putative; PR; PR-A; PR-B; Progesterone receptor; progesterone receptor A; progesterone-receptor; putative form B; steroid hormone receptor; testicular progesterone receptor; unnamed protein product | |
Rabbit | |
Protein A | |
RUO | |
5241 | |
-20° C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |