missing translation for 'onlineSavingsMsg'
Learn More

Proglumide sodium salt, Tocris Bioscience™

Product Code. 18024382
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-selective CCK antagonist; orally active

Non-selective cholecystokinin (CCK) antagonist. Inhibits CCK-stimulated amylase secretion and prevents CCK-induced 2-deoxyglucose uptake in mouse pancreatic acini. Blocks growth of HT29 colon carcinoma cells in response to gastrin 17 treatment. Orally active.

Specifications

CAS 99247-33-3
Molecular Formula C18H25N2NaO4
Quantity 50 mg
Synonym proglumide sodium, proglumide sodium salt, dl-4-benzamido-n,n-dipropylglutaramic acid sodium salt, glutaramic acid, 4-benzamido-n,n-dipropyl-, sodium salt, dl, pentanoic acid, 4-benzoylamino-5-dipropylamino-5-oxo-, monosodium salt, +-, 4-benzoylamino-5-dipropylamino-5-oxopentanoic acid sodium-potassium salt, sodium 4-dipropylcarbamoyl-4-phenylformamido butanoate, n-benzoyl-n',n'-dipropyl-dl-isoglutamine sodium-potassium salt, proglumide,sodium salt, opera_id_1870
InChI Key UFMWGCINVOIJSO-UHFFFAOYSA-M
SMILES CCCN(CCC)C(=O)C(CCC(=O)[O-])NC(=O)C1=CC=CC=C1.[Na+]
IUPAC Name sodium;4-benzamido-5-(dipropylamino)-5-oxopentanoate
Molecular Weight (g/mol) 356.398
PubChem CID 23677833
Formula Weight 356.39
Percent Purity >99%
Chemical Name or Material Proglumide sodium salt
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.