missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Proline Plus Mechanical Pipette Multipacks
GREENER_CHOICEBIOPHARMA

Multipacks combine a set of Proline Plus single-channel mechanical pipettes with matching tip trays and useful accessories. Sartorius Proline Pus Mechanical Pipette Multipacks provides best alternative to replace your used mechanical pipettes.

Brand:  Sartorius LH-728677

562.99 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.


 View more versions of this product

Product Code. 15613104

  • £720.00 / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: 3 MAX single-channel pipets (100 to 1000μL, 500 to 5000μL, 1 to 10mL), Tip Racks to 96 tips (10 to 1000μL), Bulk in a Bag: 0.1 to 5mL (100 tips), 1 to 10mL (250 tips), 1 x Linear Stand and 3 x Pipet Holders
This item is not returnable. View return policy

Description

Description

  • Low pipetting forces
  • Ergonomic finger hook
  • Easy volume setting with click stop mechanism
  • Quick and easy-to-read volume display minimises risk of volume setting errors
  • Thermal insulation of internal components improves accuracy
  • Easy to use 'do-it-yourself' calibration
  • Easy to clean with only three parts to disassemble
  • Fully autoclavable
  • High chemical and UV-resistance
  • High-quality standard Optifit tips for various applications
  • Optiload tip loading mechanism allows tip loading with an equal constant force onto every channel and enables perfect tip sealing onto every individual tip cone

Services

Two years

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Optifit Tips
Reusable
1
Non-sterile
100 to 1000 μL, 500 to 5000 μL, 1 to 10 mL
3 MAX single-channel pipets (100 to 1000μL, 500 to 5000μL, 1 to 10mL), Tip Racks to 96 tips (10 to 1000μL), Bulk in a Bag: 0.1 to 5mL (100 tips), 1 to 10mL (250 tips), 1 x Linear Stand and 3 x Pipet Holders
UV resistant
Autoclavable
Proline Plus
Proline Plus
Mechanical Pipette Multipacks
Safe-Cone Filter
1/Pk.
2 Years
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius Proline Plus Mechanical Pipette Multipacks > Proline Plus 3-Pack Max

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.