missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProLong™ Diamond Antifade Mountant with SYTOX™ Deep Red

Product Code. 17108302
Click to view available options
Volume (Metric):
1 x 2 mL
5 x 2 mL
Product Type:
ProLong Antifade Mountant
Unit Size:
2mL
Pack of 5
This item is not returnable. View return policy

Product Code. 17108302

Brand: Invitrogen™ P36990

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ProLong Diamond Antifade Mountant is compatible with almost any fluorescent dye or fluorescent protein across visible and IR spectrums. This mountant contains SYTOX Deep Red DNA stain for staining the nuclei of fixed cells.

ProLong Diamond mounting media is the ideal option for archiving samples with fluorescent dyes or fluorescent proteins. ProLong Diamond mountant cures at 1.47 RI and enables longer-term storage of your samples, while protecting them from photobleaching. ProLong Diamond Antifade Mountant comes ready to use in a 2 mL dropper bottle with SYTOX Deep Red DNA stain for nuclear counterstaining. ProLong Diamond Antifade Mountant with SYTOX Deep Red stain has an excitation of 660 nm when bound to DNA with an emission maximum at 682 nm detected using a Cy5/deep red traditional filter. Just apply a drop to the sample, add a coverslip, cure, and image. ProLong Diamond mountant does not discolor or shrink when cured and stored long term, making it possible to take high quality fluorescent images.

ProLong Diamond Antifade Mountant is compatible with a wide array of fluorescent dyes, including Alexa Fluor dyes, FITC, TRTC, Texas Red, Cy3, and Cy5. ProLong Diamond mounting media does not quench fluorophores, and its ability to provide brighter signal, against low background, helps ensure that you get the best-quality, high resolution images for publishing.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Vial
Product Type ProLong Antifade Mountant
Content And Storage Contains 1 dropper bottle containing 2 mL reagent each

• Storage of product at -5 to -25°C once received is recommended.
• Protect from light.
• Stable for at least 6 months after receiving.

Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Product Line ProLong
Reagent Type Mounting Solution, Antifade Solution
Volume (Metric) 1 x 2 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.