missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProLong™ Gold Antifade Mountant with DNA Stain DAPI
GREENER_CHOICE

ProLong Gold Antifade Mountant with DNA Stain DAPI come mixed and ready-to-use to make counterstaining more convenient and consistent while protecting fluorescently stained samples.

Brand:  Invitrogen™ P36931

205.64 GBP valid until 2025-02-28
Use promo code "24154" to get your promotional price.


 View more versions of this product

Product Code. 11549306

  • £232.00 / 10mL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

ProLong Gold Antifade Mountant with DNA Stain DAPI is a mounting media that can be directly applied to fluorescently labeled cell or tissue samples on microscope slides. It comes ready-to-use and stored at room temperature—just apply a drop, add a coverslip, cure, and image. ProLong Gold Antifade Mountants come in a 2-mL dropper bottle or 10-mL bottle. It is formulated with the blue DNA stain DAPI to make your nuclear counterstaining more convenient and consistent when preparing your samples for imaging.

ProLong Gold Antifade Mountant with DNA Stains DAPI contains chemical components designed to protect fluorescent dyes from fading (photobleaching) during fluorescence microscopy experiments. It is a curing mounting media that forms an optical path of 1.47 RI and allows longer-term storage of the sample.

Many mountants can quench initial fluorescence signal strength by up to 80%. ProLong Gold Antifade Mountant was developed to minimize this initial signal quenching for the broadest range of colors and dyes and is formulated with the DAPI DNA nuclear stain to simplify your experiments. DAPI stain is excited by UV light at 360 nm when bound to DNA, with an emission maximum at 460 nm, and is detected using a DAPI traditional filter.

ProLong Gold Antifade Mountant is not recommended for mounting samples containing fluorescent proteins such as GFP or TagRFP but can be used with Alexa Fluor dyes of your choice. For superior antifade protection of fluorescent proteins and fluorescent dyes, ProLong Diamond or ProLong Glass Antifade Mountants are recommended. ProLong Glass mountants are recommended for all immersion oil imaging applications. For immediate viewing of a sample, choose our non-curing mountant, SlowFade Gold Antifade Mountant with DAPI.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 mL
Storage at room temperature is recommended but can also be stored frozen (-5 to -30°C). Protect from light.
Less hazardous, Sustainable packaging
Mounting Solution, Antifade Solution
ProLong™ Gold Antifade Mountant with DAPI, 1 x 10 mL
Antifade Mountant
Room Temperature
ProLong™
10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.