Learn More
Molecular Probes™ ProLong™ Live Antifade Reagent, for live cell imaging
Prolong Live Antifade Reagent helps prevent the loss of fluorescent signal (photobleaching) during live cell imaging.
£62.25 - £219.00
Specifications
Product Type | Live Antifade Reagent |
---|---|
Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
Product Line | ProLong™ |
Reagent Type | Cell Imaging Reagent |
Volume (Metric) | 1 mL |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15305979
|
Molecular Probes™
P36975 |
ProLong™ Live Antifade Reagent for live cell imaging, 1 x 1 mL |
£62.25
1mL |
In Stock
Log in to see stock. |
|||||
15421204
|
Molecular Probes™
P36974 |
ProLong™ Live Antifade Reagent for live cell imaging, 5 x 1 mL |
£219.00
Pack of 5 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||||
Description
ProLong Live Antifade Reagent helps prevent the loss of fluorescent signal due to photobleaching during live cell imaging through use of enzymes from the plasma membrane of E. coli. These enzymes metabolize the elements that can cause photobleaching. When dyes are illuminated, they can degrade which reduces the sample’s fluorescent intensity and leads to the creation of free radical singlet oxygen which degrades neighboring dye molecules. ProLong Live Antifade Reagent metabolizes this element, increasing the signal intensity and duration while keeping the background signal low. ProLong Live reagent has little-to-no effect on cellular viability, proliferation, or other cellular functions in experiments up to 48 hours. ProLong Live reagent provides photobleach protection in live cell experiments for fluorophores such as GFP, RFP, Hoechst 33342, MitoTracker™, LysoTracker™, and CellTracker™dyes. ProLong Live Antifade Reagent can be added to any cell culture media or buffer that is suitable for fluorescent imaging.
Specifications
Live Antifade Reagent | |
ProLong™ | |
1 mL |
Approved for shipment at Room Temperature or on Wet Ice | |
Cell Imaging Reagent |