missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Promethazine hydrochloride
£30.05 - £134.67
Chemical Identifiers
CAS | 58-33-3 |
---|---|
Molecular Formula | C17H20N2S·HCl |
Molecular Weight (g/mol) | 320.88 |
InChI Key | XXPDBLUZJRXNNZ-UHFFFAOYSA-N |
Synonym | promethazine hydrochloride, phenergan, promethazine hcl, fenergan, n,n-dimethyl-1-10h-phenothiazin-10-yl propan-2-amine hydrochloride, diprasine, farganesse, fellozine, pipolfen, pipolphen |
PubChem CID | 6014 |
ChEBI | CHEBI:8462 |
IUPAC Name | N,N-dimethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride |
SMILES | CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)N(C)C.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15740849
|
Thermo Scientific Chemicals
254580050 |
5 g |
£30.05
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15750849
|
Thermo Scientific Chemicals
254580250 |
25 g |
£56.98
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15760849
|
Thermo Scientific Chemicals
254581000 |
100 g |
£134.67
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
58-33-3 | |
320.88 | |
promethazine hydrochloride, phenergan, promethazine hcl, fenergan, n,n-dimethyl-1-10h-phenothiazin-10-yl propan-2-amine hydrochloride, diprasine, farganesse, fellozine, pipolfen, pipolphen | |
CHEBI:8462 | |
CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)N(C)C.Cl |
C17H20N2S·HCl | |
XXPDBLUZJRXNNZ-UHFFFAOYSA-N | |
6014 | |
N,N-dimethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride |
Specifications
58-33-3 | |
C17H20N2S·HCl | |
(10% in water) Clear colorless to light yellow | |
CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)N(C)C.Cl | |
320.88 | |
CHEBI:8462 | |
Promethazine hydrochloride |
227.0°C to 230.0°C | |
promethazine hydrochloride, phenergan, promethazine hcl, fenergan, n,n-dimethyl-1-10h-phenothiazin-10-yl propan-2-amine hydrochloride, diprasine, farganesse, fellozine, pipolfen, pipolphen | |
XXPDBLUZJRXNNZ-UHFFFAOYSA-N | |
N,N-dimethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride | |
6014 | |
320.88 |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-375-2
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Promethazine hydrochloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Promethazine hydrochloride
RUO – Research Use Only