Learn More
Thermo Scientific™ ProPac™ Elite WCX HPLC Column
Shop All Thermo Scientific ProductsBeskrivelse
This column provides consistent separations of therapeutic proteins including monoclonal antibodies, antibody drug conjugates, biosimilars and charge variants.
Discover the unique Thermo Scientific™ ProPac™ Elite WCX weak cation exchange (WCX) liquid chromatography column. Designed for protein characterization including therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs) and biosimilar proteins, this column offers high resolution, high efficiency separation of proteins and their charge variants.
The unique column chemistry provides excellent performance under a broad range of pH, temperature and mobile phase compositions with excellent recovery and low carryover.
- Superior resolution power for proteins, monoclonal antibodies, and associated charge variants
- High efficiency with reproducible separa-tions
- High recovery with low carry-over
- Wide pH operating range: 2 – 12
- High temperature stability: up to 60°C
- High throughput
- Compatible with CX-1 pH Gradient Buffers
Tekniske data
Tekniske data
| Column Format | Analytical Column |
| Diameter (Metric) | 2 mm |
| Flow Rate | 0.1 to 0.25 mL/min |
| Length (Metric) | 150 mm |
| Max. Pressure | 4500 psi (310 bar) |
| Mobile Phase Compatibility | Recommended: Goods Buffers and NaCl. Minimum 20 mM salt required. Do not rinse with pure DI H2O |
| Particle Size | 5 μm |
| pH | 2 to 12 |
| Pore Size | Nonporous |
| Quantity | 1 Column |
| Vis mere |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.