Learn More
Thermo Scientific™ ProPac™ Elite WCX HPLC Column
This column provides consistent separations of therapeutic proteins including monoclonal antibodies, antibody drug conjugates, biosimilars and charge variants.
£824.10 - £3457.20
Specifications
Column Format | Analytical Column |
---|---|
Max. Pressure | 4500 psi (310 bar) |
Mobile Phase Compatibility | Recommended: Goods Buffers and NaCl. Minimum 20 mM salt required. Do not rinse with pure DI H2O |
Particle Size | 5 μm |
pH | 2 to 12 |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
15903942
|
Thermo Scientific™
303028 |
50 mm | 2 mm | 1 Column |
£824.10
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||
15993932
|
Thermo Scientific™
303027 |
150 mm | 2 mm | 1 Column |
£1095.45
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
16356931
|
Thermo Scientific™
302973 |
50 mm | 4 mm | 1 Column |
£849.23
Each |
In Stock
Log in to see stock. |
||||
15943932
|
Thermo Scientific™
302972 |
150 mm | 4 mm | 1 Column |
£1090.43
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||
15973932
|
Thermo Scientific™
303025 |
250 mm | 4 mm | 1 Column |
£1256.25
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||
15953932
|
Thermo Scientific™
302976 |
150 mm | 4 mm | 3 Columns/1 Lot |
£3457.20
Pack of 3 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||
15963932
|
Thermo Scientific™
302977 |
150 mm | 4 mm | 3 Columns/3 Lots |
£3271.28
Pack of 3 |
Estimated Shipment: 13-06-2025
Log in to see stock. |
||||
16575934
|
Thermo Scientific™
303355 |
150 mm | 9 mm | 1 Column |
£3165.75
Each |
Estimated Shipment: 13-06-2025
Log in to see stock. |
||||
Description
Discover the unique Thermo Scientific™ ProPac™ Elite WCX weak cation exchange (WCX) liquid chromatography column. Designed for protein characterization including therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs) and biosimilar proteins, this column offers high resolution, high efficiency separation of proteins and their charge variants.
The unique column chemistry provides excellent performance under a broad range of pH, temperature and mobile phase compositions with excellent recovery and low carryover.
Product Highlights- Superior resolution power for proteins, monoclonal antibodies, and associated charge variants
- High efficiency with reproducible separa-tions
- High recovery with low carry-over
- Wide pH operating range: 2 – 12
- High temperature stability: up to 60°C
- High throughput
- Compatible with CX-1 pH Gradient Buffers
Specifications
Analytical Column | |
Recommended: Goods Buffers and NaCl. Minimum 20 mM salt required. Do not rinse with pure DI H2O | |
2 to 12 | |
10°C to 60°C | |
ProPac | |
Ion Exchange | |
Bio-LC |
4500 psi (310 bar) | |
5 μm | |
Nonporous | |
Carboxylate | |
Nonporous DVB Particles with Hydrophilic Coating | |
Ion Exchange | |
Responsibly packaged |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.