missing translation for 'onlineSavingsMsg'
Learn More
Propane-1,2-Diol, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™ Product Code.: 10346290

Propane-1,2-Diol, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™

Product Code. 10346290
500 mL, Amber glass bottle
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Amber glass bottle
Metal can
This item is not returnable. View return policy

Product Code. 10346290

Brand: Fisher Chemical P/7440/08

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Propane-1,2-Diol, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number 64272
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrostShow MoreShow Less
PubChem CID 1030
ChEBI CHEBI:16997
IUPAC Name propane-1,2-diol
SMILES CC(CO)O

Specifications

Melting Point -60°C
Boiling Point 187°C
Packaging Amber glass bottle
Quantity 500 mL
Vapor Density 2.62
Formula Weight 76.1g/mol
Vapor Pressure 0.13mbar at 20°C
Physical Form Viscous Liquid
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Propane-1,2-Diol, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.