missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Propane TECLU Burner

Brand:  Bochem™ 7410

Product Code. 10547771

  • £147.00 / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Teclu Burner
Natural Gas, Propane
76 mm
Needle Valve
1 Ea.
Rheodyne Model 7410
Nickel, Steel
165 mm
1300°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers