Learn More
Propargyl bromide, 80 wt.% solution in toluene, stabilized, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 131480500
33.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 29
Additional Details : CAS Number : 106-96-7 Weight : 0.06900kg Transport : UN number : 3286 Chem class : 3 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
Propargyl bromide | |
106-96-7,108-88-3 | |
C3H3Br | |
06,493; 10,332; 06,495 | |
propargyl bromide, 3-bromopropyne, 3-bromo-1-propyne, 1-propyne, 3-bromo, 2-propynyl bromide, propynyl bromide, 1-bromo-2-propyne, propyne, 3-bromo, gamma-bromoallylene, 1-brom-2-propin | |
YORCIIVHUBAYBQ-UHFFFAOYSA-N | |
3-bromoprop-1-yne | |
7842 | |
73 to 87% (propargyl bromide) (GC) 13 to 27% (toluene) (GC) | |
1.4900 to 1.4960 | |
50 mL | |
Powder |
White | |
88.0°C to 90.0°C | |
HC≡CCH2Br | |
1.3800g/mL | |
Solubility in water: immiscible. | |
C#CCBr | |
118.961 | |
118.96 | |
Glass bottle | |
1.38 | |
4°C |
Safety and Handling
- Propargyl bromide, 80 wt% solution in toluene, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H361-Suspected of damaging fertility or the unborn child.
- H373-May cause damage to organs.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
RUO – Research Use Only