Learn More
Bochem™ Propeller Stirrers
Mixes media with low and middle viscosity
Brand:  Bochem™ 9114
Product Code. 10449582
  • £31.30 / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Three fixed blades
  • Rpm-range middle
  • Mixing of media with low and middle viscosity
  • Ideal for homogenising and suspensioning
  • Axial flow
Specifications
Specifications
Stainless Steel (18/8)
Ideal for Homogenizing and Suspensieren
1/Pk.
Propeller Stirrers
500 mm