missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Propidium Iodide ReadyProbes™ Reagent
Propidium Iodide ReadyProbes™ Reagent
Brand: Molecular Probes™ R37108
Code : NEW
Additional Details : Weight : 0.03000kg
Description
Propidium iodide is a commonly used dead-cell stain that emits red fluorescence when bound to DNA. Propidium iodide is also used as a counterstain in multicolor fluorescent techniques. With Propidium Iodide ReadyProbes™ Reagent, we provide a room temperature-stable solution of classic DNA stain that comes in a convenient-to-use dropper bottle. Just tip and drip two drops per ml to stain your cells.Features of Propidium Iodide ReadyProbes™ Reagent:
Classic Dead Cell Stain
Propidium iodide is a classic fluorescent dye used extensively for staining of dead cells (Figure 1). It exhibits a sufficiently large Stokes shift to allow simultaneous detection of nuclear DNA and FITC or Alexa Fluor™ 488-labeled antibodies.
Packaged for Convenience
Propidium Iodide ReadyProbes™ Reagent is supplied ready-to-use and can be stored right next to your flow cytometer or in your cell culture lab. A liquid formulation in an ultra-convenient dropper bottle (Figure 2) allows you to stain your cells whenever and wherever they are ready. Just add two drops per ml for optimally bright and distinct nuclear counter-staining.
Multiple Applications
Propidium iodide is membrane-impermeant and excluded from viable cells. It is therefore commonly used for identifying dead cells in a population. Its spectral properties, selectivity for dsDNA, and increased fluorescence when bound to DNA, make propidium iodide ideal for use with green (Alexa Fluor™ 488, FITC, GFP) fluorophores in multicolor experiments. Because of its high affinity for DNA, it is also frequently used for counting cells, measuring apoptosis, and sorting cells based on DNA content.
Suggestions for Use
See other Molecular Probes™ imaging tools and reagents.
Specifications
Cell-impermeant | |
6 x 2.5 mL dropper bottles Store at ≤ 25°C |
|
Viability Assay | |
Nucleic Acid Stain | |
617 nm | |
Liquid | |
6 Bottles |
Far-Red | |
Fluorescence | |
Attune Acoustic Focusing Cytometer, EVOS Cell Imaging System, Flow Cytometer | |
propidium iodide | |
535 nm | |
ReadyProbes | |
Nucleus |
For Research Use Only. Not for use in diagnostic procedures.