missing translation for 'onlineSavingsMsg'
Learn More

Propyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10634025
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10634025

Brand: Thermo Scientific Maybridge TL01934DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 628-30-8
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.167
MDL Number MFCD00004823
InChI Key KKASGUHLXWAKEZ-UHFFFAOYSA-N
Synonym propyl isothiocyanate, propylisothiocyanate, propane, 1-isothiocyanato, n-propyl isothiocyanate, isothiocyanic acid propyl ester, isothiocyanic acid, propyl ester, isothiocyanic acid n-propyl ester, 1-propyl isothiocyanate, propanisothiocyanate, 1-isothiocyanatopropan
PubChem CID 69403
IUPAC Name 1-isothiocyanatopropane
SMILES CCCN=C=S

Specifications

CAS Min % 97.0
Density 0.9780g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3CH2CH2NCS
Quantity 1 g
Formula Weight 101.17
Percent Purity 97%
Chemical Name or Material Propyl isothiocyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.