missing translation for 'onlineSavingsMsg'
Learn More

Propyl propionate, 98+%

Product Code. 10753614 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
This item is not returnable. View return policy

Product Code. 10753614

Brand: Thermo Scientific Acros 385520010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-36-5 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-36-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009373
InChI Key MCSINKKTEDDPNK-UHFFFAOYSA-N
Synonym propyl propionate, n-propyl propionate, propanoic acid, propyl ester, n-propyl propanoate, propionic acid, propyl ester, n-propyl n-propionate, propylester kyseliny propionove, unii-g09trv00gk, propyl propionate natural, propionic acid n-propyl ester
PubChem CID 7803
IUPAC Name propyl propanoate
SMILES CCCOC(=O)CC

Specifications

Melting Point -76.0°C
Color Colorless
Density 0.8810g/mL
Boiling Point 123.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2C(O)O(CH2)2CH3
Refractive Index 1.3820 to 1.3940
Quantity 1 L
Merck Index 15, 7979
Specific Gravity 0.881
Solubility Information Solubility in water: soluble
Formula Weight 116.16
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Propyl propionate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.