Learn More
Propylamine, 99+%, extra pure, Thermo Scientific Chemicals
£15.20 - £78.10
Chemical Identifiers
CAS | 107-10-8 |
---|---|
Molecular Formula | C3H9N |
Molecular Weight (g/mol) | 59.11 |
MDL Number | MFCD00008205 |
InChI Key | WGYKZJWCGVVSQN-UHFFFAOYSA-N |
Synonym | propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine |
PubChem CID | 7852 |
ChEBI | CHEBI:39870 |
IUPAC Name | propan-1-amine |
SMILES | CCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10723031
|
Thermo Scientific Acros
220610500 |
50 mL | Glass bottle |
£15.20
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10697192
|
Thermo Scientific Acros
220615000 |
500 mL | Glass bottle |
£25.50
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10052920
|
Thermo Scientific Acros
220610010 |
1 L | Glass bottle |
£37.90
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13418478
|
Thermo Scientific Acros
220610025 |
2.5 L | Glass bottle |
£78.10
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-10-8 | |
59.11 | |
WGYKZJWCGVVSQN-UHFFFAOYSA-N | |
7852 | |
propan-1-amine |
C3H9N | |
MFCD00008205 | |
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine | |
CHEBI:39870 | |
CCCN |
Specifications
107-10-8 | |
0.7190g/mL | |
−30°C | |
99% min. (GC) | |
CH3CH2CH2NH2 | |
MFCD00008205 | |
13,164 | |
0.719 | |
Solubility in water: soluble in water. Other solubilities: miscible with alcohol and ether | |
CCCN | |
59.11 | |
CHEBI:39870 | |
59.11 | |
Extra Pure | |
Propylamine, 99+% |
-83.0°C | |
48.0°C | |
Authentic | |
C3H9N | |
1.3870 to 1.3890 | |
04,136 | |
15,7956 | |
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine | |
WGYKZJWCGVVSQN-UHFFFAOYSA-N | |
propan-1-amine | |
7852 | |
0.4 mPa.s (20°C) | |
99+% | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-462-3
missing translation for 'rtecsNumber' : UH9100000
missing translation for 'tsca' : TSCA
- Propylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only