missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylene Glycol, MP Biomedicals
£45.95 - £65.40
Chemical Identifiers
CAS | 57-55-6 |
---|---|
Molecular Formula | C3H8O2 |
Molecular Weight (g/mol) | 76.095 |
InChI Key | DNIAPMSPPWPWGF-UHFFFAOYSA-N |
Synonym | 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost |
PubChem CID | 1030 |
ChEBI | CHEBI:16997 |
IUPAC Name | propane-1,2-diol |
SMILES | CC(CO)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11407232
|
MP Biomedicals
0215195790 |
500 mL |
£45.95
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11417232
|
MP Biomedicals
0215195791 |
1000 mL |
£65.40
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
57-55-6 | |
76.095 | |
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
CHEBI:16997 | |
CC(CO)O |
C3H8O2 | |
DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
1030 | |
propane-1,2-diol |
Specifications
57-55-6 | |
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
CC(CO)O | |
76.095 | |
CHEBI:16997 | |
Propylene Glycol |
C3H8O2 | |
DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
propane-1,2-diol | |
1030 | |
76.09 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title