Learn More
Contec™ PROSAT™ Knit Polyester Wipes Presaturated
100% interlock knit standard weight textured polyester presaturated with 70% USP grade IPA and 30% DI water
Brand: Contec™ PSCS0009SZ
Additional Details : CAS Number : 67-63-0 Weight : 10.40000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Description
- Polywipe-C Heatseal Wipes are made from a standard weight 100% knitted polyester that is laser cut to bond the fibers at the edges of the wipe.
- The wipe is chemical resistant and exceptionally low in particles and extractable residue making it ideal for critical cleaning.
- The wipes are laundered and packed in an ISO Class 4 cleanroom.
- These wipes are presaturated with 70% USP grade isopropanol and 30% DI water, presaturation greatly reduces solvent usage and reduces the VOCs associated with spraying IPA.
- Using presaturated wipes also maximizes process control and improves consistency between operators.
- The wipes are gamma irradiated and validated per AAMI Guidelines to a 10-6 SAL, so can be used in the highest grade life science cleanrooms.
- The wipes are provided in convenient and easy to use peel and reseal pouches.
- The resealable pouch with the option of a small number of wipes per pouch helps reduce waste and control saturation volume and cleanliness.
- Features & Benefits:
- Recommended for Grade A/B or ISO Class 3-8 environments
- Standard weight laundered knitted 100% polyester fabric for extremely low levels of particles and fibers
- Heatsealed edges prevent fiber generation during use
- Small number of wipes per resealable pouch helps eliminates waste
- Presaturated wipe reduces VOC emissions and solvent use
- Resealable pouch preserves cleanliness and solvent saturation levels
Specifications
Wipe | |
23 cm | |
23 cm | |
9 x 9 in. | |
70% IPA/ 30% DI Water |
9 in. | |
9 in. | |
Heat-sealed edges | |
24 Pouches/Cs. | |
Sterile |