Learn More
Contec™ PROSAT™ Presaturated Knitted Polynit Wipes
Clean surfaces and equipment in critical environments where aggressive solvents are needed. The PROSAT™ Presaturated Knitted Polynit Wipes are pre-moistened polyester wipes for general purpose cleanings.
£333.00 - £937.00
Specifications
Product Type | Pre-saturated Wipe |
---|---|
Material | Polyester |
Autoclavable | Not Autoclavable |
Basis Weight | 140 gsm |
Construction | No-run pique knit |
Product Code | Brand | Dimensions (L x W) | Saturation Solution | Sterility | Packaging Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Saturation Solution | Sterility | Packaging Quantity | Price | Quantity & Availability | ||
17274523
|
Contec™
PSCS0009-BPR |
23 x 23 cm (9 x 9 in.) | 70% IPA, 30% DI Water | Sterile | 20 Wipes/Pouch, 24 Pouches/Cs. (Folded) |
£455.00
Pack of 480 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17294513
|
Contec™
PS-HS9-7030-BPR |
23 x 23 cm (9 x 9 in.) | 70% IPA, 30% DI Water | Non-sterile | 30 Wipes/Pouch, 30 Pouches/Cs. (Flat Stacked) |
£835.00
Pack of 900 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17224533
|
Contec™
PSPS0076-BPR |
23 x 23 cm (9 x 9 in.) | 70% IPA, 30% DI Water | Sterile | 20/Pk., 5 Pk./OB1, 1 OB1/OB2, 8 OB2/Cs. |
£937.00
Pack of 800 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17244533
|
Contec™
PSPS0091-BPR |
30 x 30 cm (12 x 12 in.) | 70% IPA, 30% DI Water | Sterile | 20 Wipes/Pouch, 20 Pouches/Cs. (Flat Stacked) |
£823.00
Pack of 400 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16585274
|
Contec™
PSPS0044 |
10 x 10 cm (4 x 4 in.) | 70% IPA, 30% DI Water | Sterile | 20 Wipes/Pouch, 64 Pouches/Cs. |
£917.00
Pack of 160 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15940502
|
Contec™
PS-HS9-7030 |
23 x 23 cm (9 x 9 in.) | 70% IPA, 30% DI Water | Non-sterile | 30 Wipes/Pouch, 30 Pouches/Cs. | N/A | |||
16645534
|
Contec™
PSCS0009 |
23 x 23 cm (9 x 9 in.) | 70% IPA, 30% DI Water | Sterile | 20 Wipes/Pouch, 24 Pouches/Cs. | N/A | |||
15674937
|
Contec™
PSPS0093 |
23 x 23 cm (9 x 9 in.) | 70% IPA, 30% DI Water | Sterile | 20 Wipes/Pouch, 40 Pouches/Cs. (Half-folded) | N/A | |||
15197915
|
Contec™
PSPS0076 |
23 x 23 cm (9 x 9 in.) | 70% IPA, 30% DI Water | Sterile | 20 Wipes/Pouch, 40 Pouches/Cs. | N/A | |||
15534045
|
Contec™
PSPS0091 |
30 x 30 cm (12 x 12 in.) | 70% IPA, 30% DI Water | Sterile | 20 Wipes/Pouch, 20 Pouches/Cs. | N/A | |||
16459579
|
Contec™
PSPS0047 |
30 x 30 cm (12 x 12 in.) | 70% IPA, 30% DI Water | Non-sterile | 30 Wipes/Pouch, 20 Pouches/Cs. |
£741.00
Pack of 600 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15187915
|
Contec™
PSPS0073 |
15 x 23 cm (6 x 9 in.) | 70% IPA, 30% DI Water | Sterile | 30/Pouch, 20 Pouches/Cs. | N/A | |||
16675534
|
Contec™
PSPS0077 |
23 x 23 cm (9 x 9 in.) | 70% IPA, 30% DI Water | Sterile | 20 Wipes/Pouch, 40 Pouches/Cs. | N/A | |||
Description
Clean surfaces and equipment in critical environments where aggressive solvents are needed. The Contec™ PROSAT™ Presaturated Knitted Polynit Wipes are premoistened polyester wipes for general purpose cleanings. Wipes are textured with a no-run interlock knit for good solvent sorbency, abrasion resistance and chemical resistance.
- Ideal for applications where large quantities of wipes or aggressive solvents are needed for wiping surfaces and equipment.
- Available with both 100% Semi Grade IPA and 70% USP Grade IPA, 30% DI Water.
- Extremely low in particle and fiber generation.
- Soft, non-abrasive textured wipe cleans without scratching.
- Premoistened polyester wipes are designed for general-purpose cleaning in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments.
- Ideal for wiping exterior product surfaces and cleaning the interior surfaces of process tools or other equipment.
- Standard weight wipes are made of 100% textured polyester with no-run interlock knit for good solvent sorbency, abrasion resistance, and chemical resistance.
- Heatsealed and saturated with various blends of isopropyl alcohol and deionized water.
- Low in particles and extractables.
- Laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
- Sterile wipes are Validated Sterile to 10-6 Sterility Assurance level in accordance with AAMI/ISO 11137 Method 1 Guidelines for the sterilization of medical devices.
- Sterile PROSAT™ Knitted Polynit Wipes are conveniently packaged for use in sterile core areas and are ideal for wiping articles prior to pass-through and environmental cleaning.
Specifications
Pre-saturated Wipe | |
Not Autoclavable | |
No-run pique knit | |
1-ply |
Polyester | |
140 gsm | |
Single-use |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.