Learn More
Contec™ PROSAT™ Presaturated Nonwoven Wipes

Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.
Brand: Contec™ PS-850-BPR
530.47 GBP valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. Contec™ PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.
- Wipe substrate designed for the application
- Compatible with ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
- Ideal for wiping articles prior to pass through
- Made of meltblown polypropylene or cellulose/polyester
- Effective and convenient for wipe-downs of process tools and other equipment
- Choose spunlaced 100% polyester wipers for improved cleanliness, meltblown polypropylene wipes for improved durability, or economical hydroentangled polyester/cellulose wipers
- Solvent blend meets SEMI specifications for metals and contaminants

Specifications
Pre-saturated Wipe | |
20 cm | |
20 cm | |
Not Autoclavable | |
Single-use | |
Ideal for Routine Cleaning and Wiping Instruments and Equipment | |
50 Pk., 50 Pk./Cs. | |
70% IPA, 30% DI Water |
8 in. | |
8 in. | |
Polypropylene | |
ISO Class 5 | |
20 x 20 cm (8 x 8 in.) | |
Process Tools and Other Equipment | |
1-ply | |
Non-sterile |
Safety and Handling
- CONTEC PROSAT MBPP 70/30 IPA & DI WATER PS-850
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1092463-0-0-1 | Certificate of Quality | 01/03/2025 | PS-850-BPR |
1092462-0-0-1 | Certificate of Quality | 17/01/2025 | PS-850-BPR |
1089631-0-0-1 | Certificate of Quality | 10/01/2025 | PS-850-BPR |
1079458-0-0-1 | Certificate of Quality | 17/10/2024 | PS-850-BPR |
1075146-0-0-1 | Certificate of Quality | 20/09/2024 | PS-850-BPR |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Days
33Hours
0Minutes
7Seconds
33New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.