Learn More
Contec™ PROSAT™ PS-911 Presaturated Meltblown Polypropylene Wipes
PROSAT™ wipes are an effective, efficient and convenient method of cleaning process tools and other equipment. PROSAT wipes are also ideal for wiping articles prior to pass through.
Marke: Contec™ PS-911-BPR
427.00 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Weitere Details : Weight : 11.59200kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Beschreibung
- Presaturated with IPA and DI water blends in flat, resealable pouches individually lot-numbered for traceability
- These specially designed pouches preserve cleanliness and solvent saturation levels
- Solvent blend meets SEMI specifications for metals and contaminants
- ISO Class 5 compatible
- 30 wipes/pouch, 50 pouches/case
Spezifikation
Pre-saturated Wipe | |
23 cm | |
28 cm | |
ISO Class 5, SEMI for metals/contaminants | |
23 x 28 cm (9 x 11 in.) | |
Process Tools and Other Equipment | |
70% IPA/ 30% DI Water |
9 in. | |
11 in. | |
Polypropylene | |
Single-use | |
Ideal for Routine Cleaning and Wiping Instruments and Equipment | |
30 Pk., 50 Pk./Cs. | |
Non-sterile |
Sicherheit und Handhabung
- CONTEC PROSAT MBPP WITH 70% IPA
Signalwort
- Danger
Gefahrenkategorie
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Gefahrenhinweis
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Sicherheitshinweis
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Ergänzende Informationen
- MIXTURE LIST-Contains : propan-2-ol