missing translation for 'onlineSavingsMsg'
Learn More

Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes

Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.

Brand:  Contec™ PSC20001

Code : NEW

Additional Details : CAS Number : 67-63-0 Weight : 0.33300kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 11917025

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.

  • Designed for pharmaceutical, biotech, and medical device applications
  • Choose spunlaced nonwoven 100% polyester wipes for improved cleanliness, economical hydroentangled polyester/cellulose wipers, or meltblown polypropylene wipes for improved durability
  • Conveniently presaturated with 70/30 blend of semi-conductor grade IPA and DI water to reduce VOC emissions and hazardous waste
  • All wipes are sterilized by gamma radiation to a 10-6 Sterility Assurance Level and validated per AAMI ISO 11137 (Method 1) guidelines
  • Certificate of Sterility and Certificate of Irradiation included
  • Environment: ISO 5-8, Grade A/B
  • Individually lot numbered resealable pouches with tamper-evident labels ensure product integrity and cleanliness
Specifications

Specifications

PROSAT™ Pre-saturated Wipes
23 cm
27.9 cm
ISO Class 5-8
Ideal for routine cleaning and wipe down of lab tools, instruments and other equipment and designed for sterile environments
70% IPA, 30% DI Water
9 in.
11 in.
Polyester Cellulose
27.9 x 23 cm (11 x 9 in.)
24 Pk.
Sterile
Sugerencias de productos

Sugerencias de productos

Videos
Seguridad y manipulación

Seguridad y manipulación

Nombre del producto
  • CONTEC PROSAT AMP SIGMA 70/30 IPA & DI WATER

Palabra de advertencia
  • Danger

Clasificación
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Indicaciones de peligro
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.

Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P370+P378-In case of fire: Use dry chemical, carbondioxide to extinguish.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Otros peligros
  • MIXTURE LIST-Contains : PROPAN-2-OL

SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales