missing translation for 'onlineSavingsMsg'
Learn More

Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes
BIOPHARMA

Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.

Brand:  Contec™ PSC20005-BPR

525.99 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.



 View more versions of this product

Product Code. 17244523

  • £901.00 / Pack of 1400

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.

  • Designed for pharmaceutical, biotech, and medical device applications
  • Choose spunlaced nonwoven 100% polyester wipes for improved cleanliness, economical hydroentangled polyester/cellulose wipers, or meltblown polypropylene wipes for improved durability
  • Conveniently presaturated with 70/30 blend of semi-conductor grade IPA and DI water to reduce VOC emissions and hazardous waste
  • All wipes are sterilized by gamma radiation to a 10-6 Sterility Assurance Level and validated per AAMI ISO 11137 (Method 1) guidelines
  • Certificate of Sterility and Certificate of Irradiation included
  • Recommended for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
  • Individually lot numbered resealable pouches with tamper-evident labels ensure product integrity and cleanliness

Compliance

Validated sterile to a Sterility Assurance Level of 10-6 in accordance with AAMI/ISO 1137 Method 1 Guidelines.

Specifications

Specifications

Pre-saturated Wipe
23 cm
27.9 cm
ISO Class 4
Ideal for Routine Cleaning and Wipe Down of Lab Tools, Instruments and Other Equipment and Designed for Sterile Environments
70% USP grade IPA, 30% DI water
9 in.
11 in.
Polyester Cellulose
23 x 28 cm (9 x 11 in.)
50 Pk.
Sterile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • CONTEC PROSAT STERILE AMP THETA 70/30 IPA & DI WATER

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use media other than water to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.