missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 60
223.73 GBP valid until 2024-12-31
Use promo code "22918" to get your promotional price.
Use promo code "22918" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Constructed of a microporous film on a polypropylene nonwoven fabric.
- Combine good particle protection with high liquid repellency.
- Help provide a barrier against non-hazardous dry particles down to one micron in size.
- The material is permeable to moisture vapour only and is not air permeable.
- Certified according to Regulation (EU) 2016/425.
- Chemical protective clothing, Category III, Type 5 and 6.
- EN 1073-2 (protection against radioactive contamination).
- Antistatic treatment (EN 1149-5) on inside.
- Stitched internal seams.
- Nylon zipper with flap.
- Ideal choice for applications that are less demanding in terms of barrier, durability and comfort.
Specifications
Size | Small |
Gender | Unisex |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Closure Type | Zipper with Storm Flap |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'