Learn More
DuPont™ ProShield™ 60
Product Code. 16284511 Shop All DuPont Products
Click to view available options
Size:
3X-Large
Unit Size:
Pack of 50
Product Code. 16284511
Brand: DuPont™ D15519557
Only null left
Add to basket
Add to basket
ProShield™ 60 provides a barrier against non-hazardous dry particles down to one micron in size.
  • Constructed of a microporous film on a polypropylene nonwoven fabric.
  • Combine good particle protection with high liquid repellency.
  • Help provide a barrier against non-hazardous dry particles down to one micron in size.
  • The material is permeable to moisture vapour only and is not air permeable.
  • Certified according to Regulation (EU) 2016/425.
  • Chemical protective clothing, Category III, Type 5 and 6.
  • EN 1073-2 (protection against radioactive contamination).
  • Antistatic treatment (EN 1149-5) on inside.
  • Stitched internal seams.
  • Nylon zipper with flap.
  • Ideal choice for applications that are less demanding in terms of barrier, durability and comfort.
TRUSTED_SUSTAINABILITY
Specifications
Size 3X-Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Description Individually Packed
Quantity 50/Cs.