Learn More
Thermo Scientific™ ProSpecT™ C. difficile Toxin A/B Microplate Assay
Rapidly detect Clostridium difficile toxin A and B from human fecal samples.
Brand: Thermo Scientific™ R244596
1317.43 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Rapidly detect Clostridium difficile toxin A and B in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ C. difficile Toxin A/B Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with the first C. difficile Toxin A/B assay to combine excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.- Excellent clinical performance
- Cary Blair transport option which saves a dilution step
- Results within 100 minutes
- Room temperature incubation
- True colorless negatives
ProSpecT C. difficile Toxin A/B is a direct, qualitative enzyme immmunoassay for the detection of Clostridium difficile toxin A and B and is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures, and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT C. difficile Toxin A/B provides same-day toxin detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C | |
ProSpecT C. difficile Toxin A/B Microplate Assay | |
Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card | |
96 Wells/Kit |