missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ProSpecT™ STEC (Shiga Toxin E. coli) Microplate Assay

Rapidly detect E. coli Shiga Toxin 1 and 2 from human fecal samples.

£935.00

Specifications

Content And Storage 2°C to 8°C
Description ProSpecT STEC (Shiga Toxin E.coli) Microplate Assay
Detectable Analytes E. coli Shiga Toxin 1 and 2
Type E. coli Testing Kit
For Use With (Application) Enzyme Immunoassay Detection
View More Specs

Products 1
Product Code Brand Description Quantity Price Quantity & Availability  
Product Code Brand Description Quantity Price Quantity & Availability  
11903622
View Documents
Thermo Scientific™
R2474096
ProSpecT STEC (Shiga Toxin E.coli) Microplate Assay 96 Wells/Kit This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Rapidly detect Shiga Toxin-producing Escherichia coli in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ STEC (Shiga Toxin E. coli) Microplate Assay.

Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Shiga Toxin-producing Escherichia coli. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT STEC (Shiga Toxin E.coli) is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT STEC provides same-day toxin detection that is easy-to-use, flexible and provides the power to do more with less.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications

Specifications

2°C to 8°C
E. coli Shiga Toxin 1 and 2
Enzyme Immunoassay Detection
ProSpecT STEC (Shiga Toxin E.coli) Microplate Assay
E. coli Testing Kit
Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.

Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.