missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Prostaglandin E2

Prostanoid pathway activator; Activates prostaglandin receptors EP1, EP2, EP3 and EP4

£109.35 - £166.05

Specifications

CAS 363-24-6
Molecular Formula C20H32O5
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17110042
View Documents Promotion Details
STEMCELL Technologies
72194
5 mg
£166.05
5mg
View alternative products
Save Up to 
»
Estimated Shipment: 31-07-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15692517
View Documents Promotion Details
STEMCELL Technologies
72192
1 mg
£109.35
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 12-08-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Prostaglandin E2 (PGE2) is the most biologically active and well-studied prostaglandin
  • It binds with very high affinity to the EP1, EP2, EP3, and EP4 receptors (Ki = 9.1, 4.9, 0.33, 0.79 nM respectively)
  • (Abramovitz et al.), Bos et al.)
  • Required for the development of hematopoietic stem cells (HSCs) in mice and zebrafish (North et al.)
  • Improves engraftment of mouse HSCs, possibly through increasing homing, survival, and/or self-renewal (Hoggatt et al. 2009, Hoggatt et al.) 2013, North et al.)
  • Promotes differentiation of hematopoietic progenitor cells from mouse, macaque, and human embryonic stem cells (Gori et al.), North et al.), Woods et al.)
  • Promotes differentiation of myeloid-derived suppressor cells from hematopoietic progenitors (Sinha et al.)
  • Promotes differentiation of Th17 cells from naïve T-cells (Boniface et al.)
Specifications

Specifications

363-24-6
C20H32O5
Videos
SDS
Documents

Documents

Product Certifications