missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Prostaglandin E2 Biotrak™ Assay
Enzyme immunoassay system suitable for plasma, urine, or cell culture supernatant samples
Brand: Cytiva RPN222
Code : GH
Additional Details : Weight : 0.70000kg
Includes: RPN222 supplied with sufficient quantities of reagents for 96 wells; RPN22210, for 960 wells. RPN22210 does not include lysis reagents. Both allow for construction of one standard curve and measurement of 37 unknowns in duplicate per plate.
Description
- Specific for PGE2
- Flexible method with choice of five assay protocols
- Product available with or without lysis reagents
- Lysis reagents facilitate extraction of PGEsubscript 2 from cell cultures and plasma samples; PGE is directly available for subsequent analysis without removal of extraction reagents
- Precise and accurate measurement
- Ready to use substrate and color-coded reagents
- Range: 20-640pg/mL (1-32pg/well)
- Room temperature protocol: 50-6400pg/mL
- Sensitivity: 16pg/mL, 0.8pg/well
- 5-hour protocol
- Storage: Store at -15° to -30°C.
Specifications
20 to 640pg/mL | |
96 Wells | |
50 μL | |
5 hr. | |
ELISA |
Microplate (12 x 8-well strip coated with sheep Anti-Mouse IgG), Assay Buffer, standard (prostaglandin E2) 256ng/mL, prostaglandin E Antibody (lyophilized), Prostaglandin E2 Conjugate, Wash Buffer, TMB Substrate, lysis reagent 1, lysis reagent 2 | |
Plasma (EDTA, Citrate), Urine, Tissue Homogenates, Cell Culture, Cell Culture Supernatant | |
16 pg/mL (0.8 pg/Well) | |
-15°C to -30°C | |
Prostaglandin E2 Direct Biotrak assay |
Safety and Handling
Signal Word
- Danger