missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Prostratin, 99+%

Product Code. p-7070069
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 15413649

Brand: Thermo Scientific Chemicals J60971.M^

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

12-Deoxyphorbol 13-acetate; activator of protein kinase C. A weak mouse ear irritant, but lethal at 1ug per mouse

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60857-08-1
Molecular Formula C22H30O6

Specifications

Quantity 1 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) 12-Deoxyphorbol 13-acetate; activator of protein kinase C. A weak mouse ear irritant, but lethal at 1μg per mouse
Formula Weight 390.47
Melting Point 216°C to 219°C
Solubility Soluble in DMSO at 30mg/ml
Form Powder
InChI Key BOJKFRKNLSCGHY-KKFGZBDXSA-N
SMILES C[C@@H]1CC2(OC(C)=O)C([C@@H]3C=C(CO)C[C@@]4(O)[C@@H](C=C(C)C4=O)[C@@]13O)C2(C)C
IUPAC Name (1R,2S,6R,10S,15R)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-13-yl acetate
Molecular Weight (g/mol) 390.48
Assay 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.