Learn More
Honeywell™ PROTECT (i)XTREM S3 Shoes
Honeywell™ PROTECT (i)XTREM S3 Shoes are constructed of water-resistant full grain leather.
Brand:  Honeywell™ 6246204-48/7
 View more versions of this product
Product Code. 15826595
  • £130.00 / 1 pair
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Water resistant grain full leather
  • Ergonomic collar and padded tongue for an optimized comfort
  • Gusset to avoid water and dust penetration
  • Breathable 3D lining
  • Derby style for a better fitting in all sizes
  • Toe cap metal free
  • Metal free antiperforation insert
  • Injected PU / Nitrile sole
  • (i)NTELLIGENT wear control
  • EN ISO 20345:2011 S3 HRO HI CI SRC
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN ISO 20345:2011 S3 HRO HI CI SRC
Leather
Polyurethane and nitrile
Black
48
Metal-free