Learn More
Eppendorf™ Protection Filter Set
Designed for use with Eppendorf Reference™ 2, Eppendorf Research™ plus and Eppendorf Xplorer™ (plus) pipettes
Brand:  Eppendorf™ 4920622009
Code : NEW
Additional Details : Weight : 0.01100kg
Product Code. 15244436
  • £14.50 / 1 set
In Stock
to see stock.
Description
Description
  • Available in a variety of colors
  • Protection Filter Sets include 10 filters and 1 filter sleeve
  • For fixed- and variable-volume pipettes