Learn More
Heidolph Instruments Guard Hood
Protective cover
Brand: Heidolph Instruments
Additional Details : Weight : 1.44000kg
Description
- Strong and transparent guard hood to protect you from implosions
- Provides easy and immediate access to flask and bath
- Application range up to 130°C
Order Info
dimType
Specifications
Polycarbonate | |
Protective Cover | |
1 Ea. | |
115/230 VAC |