missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Protective Coveralls
Click to view available options
Size: XL; Color: Grey
Description
- Available in a range of sizes
- Extra comfort for extended wear
- Provides barrier protection against light liquid splashes and hazardous dust
Specifications
Product Type | Protective Coveralls |
Size | X-Large |
Material | Polyethylene |
Color | Gray |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Closure Type | Zipper |
Resistance | Liquid |
Back Style | Large Zip |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'