missing translation for 'onlineSavingsMsg'
Learn More

Mutexil™ Protective Overshoe Coverings

Product Code. 11775433
Click to view available options
Product Type:
Overshoe
Protective CPE Visitor Overshoes
Sticky Overshoe
Color:
Blue
White
Size:
2X-Large
Universal
X-Large
Unit Size:
Pack of 2000
Pack of 300

Product Code. 11775433

Brand: Mutexil™ 2000044

Only null left
Add to basket
Add to basket

Choose from a range of choices and construction, from sticky, anti-slip, fluid -resistant, polypropylene and CPE variations with Mutexil™ Protective Overshoe Coverings.

  • Various sizes and colors
  • Needs-based styles
  • Per new standards EN ISO 20345 and EN ISO 20347

Specifications

Color White
For Use With (Application) For sanitary, food, industrial, catering and services
Material Polypropylene
Size 2X-Large
Sole Features Anti-slip sole
Waterproof Yes
Product Type Sticky Overshoe
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.