Learn More
Molecular Probes™ protein A, Alexa Fluor™ 546 conjugate
protein A, Alexa Fluor™ 546 conjugate
Brand:  Molecular Probes™ P11049
Product Code. 10423632
  • £257.00 / 1mg
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
Our Alexa Fluor 546 protein A conjugate binds to the Fc portion of various immunoglobulins from several different species. This exceptionally bright, orange-fluorescent probe (absorption/emission maxima ∼556/573 nm) can be used to visualize antibodies in a manner analogous to a labeled species-specific secondary antibody.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Alexa Fluor 546
Alexa Fluor™
Protein A
Room Temperature
1 mg
Lyophilized Powder
Store in freezer (-5 to -30°C) and protect from light.