missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ Protein A–FITC Conjugate
Protein A–FITC Conjugate
Brand: Novex™ 101011
Code : 50
Additional Details : Weight : 0.50000kg
Description
Protein A (from Staphylococcus aureus, Cowan I strain) is conjugated with fluorescein isothiocyanate (FITC) and is purified by molecular sieving to remove the unbound FITC and unconjugated Protein A. The product is in 10 mM phosphate buffered saline, pH 7.4, containing 1% ovalbumin, 40% glycerol, and 0.1% sodium azide.Specifications
Fluorescein | |
Solution | |
Store at 2-8°C |
2 mL | |
Protein A | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.