Learn More
Cytiva Protein A HP SpinTrap™ Column
For simple, small-scale antibody purification or enrichment of specific proteins by antibody capture in single-use spin columns
Brand: Cytiva 28-9031-32
Code : NEW
Additional Details : Weight : 0.08820kg
Description
Prepacked with Protein A Sepharose™ High Performance Media for coupling of antibodies of IgG subclasses; used with a standard microcentrifuge
- Fast binding kinetics and high capacity provide high yield
- Classic (antigen eluted with antibody) and cross-link (antigen eluted separately) protocols provide flexibility
- Elution conditions formatted for both electrophoresis and LC-MS analysis workflows
- Easy scale-up with HiTrap Protein A HP prepacked columns
- Additional protocol for small-scale purificaiton of monoclonal and polyclonal antibodies from serum and cell culture supernatants
Specifications
>1 mg Human IgG well | |
600 μl | |
2°C to 30°C, 20% Ethanol |
Centrifugation | |
Standard microcentrifuge. | |
16 Columns |