missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Protein G, Alexa Fluor™ 488 conjugate

Product Code. 10708424
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10708424

Brand: Invitrogen™ P11065

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Protein G, Alexa Fluor™ 488 conjugate

Alexa Fluor 488 protein G conjugate exhibits bright, green fluorescence (absorption/emission ∼495/519 nm). Protein G is a bacterial protein similar to protein A but with superior selectivity for mouse and human IgG.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Alexa Fluor 488
Quantity 1 mg
Product Line Alexa Fluor
Form Lyophilized Powder
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.