Learn More
Cytiva Protein G Antibody Package Affinity Columns for Antibody Purification
Protein G is a good choice for the general purpose capture of antibodies at the laboratory scale
Brand: Cytiva 29-0588-06
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Protein G is a good choice for the general purpose capture of antibodies at the laboratory scale since it binds a broader range of IgG from eukaryotic species and binds more classes of IgG than protein A. Contents: HiTrap Protein G HP (1 x 1mL) and HiTrap Desalting (1 x 5mL)Specifications
Antibody affinity chromatography | |
2°C to 8°C | |
Complete packsize |
Safety and Handling
- Antibody package, Protein G
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.